Assay Details
| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174490.3 | NP_776915.1 | 2 - 3 | 413 | 76 | |
| GenBank mRNA | BC103271.1 | - | 2 - 3 | 413 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VSNL1| Gene Symbol: VSNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174490.3 | NP_776915.1 | 2 - 3 | 413 | 76 | |
| GenBank mRNA | BC103271.1 | - | 2 - 3 | 413 | 76 | |
Set Membership: |
|