Assay Details
| Gene Symbol: CHRNB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174517.1 | NP_776942.1 | 1 - 2 | 56 | 65 | |
| GenBank mRNA | AF487465.1 | - | 1 - 2 | 56 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHRNB4| Gene Symbol: CHRNB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174517.1 | NP_776942.1 | 1 - 2 | 56 | 65 | |
| GenBank mRNA | AF487465.1 | - | 1 - 2 | 56 | 65 | |
Set Membership: |
|