Assay Details
| Gene Symbol: CHRNB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174517.1 | NP_776942.1 | 2 - 3 | 203 | 87 | |
| GenBank mRNA | AF487465.1 | - | 2 - 3 | 203 | 87 | |
More Information
Set Membership: |
|

CHRNB4
Genomic Map