Assay Details
| Gene Symbol: GUCA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174546.2 | NP_776971.1 | 1 - 2 | 412 | 66 | |
| GenBank mRNA | X95352.1 | - | 439 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GUCA1A| Gene Symbol: GUCA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174546.2 | NP_776971.1 | 1 - 2 | 412 | 66 | |
| GenBank mRNA | X95352.1 | - | 439 | 66 | ||
Set Membership: |
|