Assay Details
| Gene Symbol: PTGER4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174589.2 | NP_777014.1 | 1 - 2 | 892 | 85 | |
| GenBank mRNA | AF539403.1 | - | 1 - 2 | 892 | 85 | |
More Information
Set Membership: |
|

PTGER4
Genomic Map