Assay Details
| Gene Symbol: PROKR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174640.1 | NP_777065.1 | 1 - 2 | 452 | 109 | |
| GenBank mRNA | AY089973.1 | - | 1 - 2 | 452 | 109 | |
More Information
Set Membership: |
|

PROKR2
Genomic Map