Assay Details
| Gene Symbol: ASB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174687.2 | NP_777112.1 | 7 - 8 | 859 | 75 | |
| GenBank mRNA | AF527382.1 | - | 7 - 8 | 859 | 75 | |
| BC123763.1 | - | 7 - 8 | 860 | 75 | ||
More Information
Set Membership: |
|

ASB15
Genomic Map