Assay Details
| Gene Symbol: PPP1R16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174824.2 | NP_777249.1 | 10 - 11 | 1384 | 61 | |
| GenBank mRNA | AF362909.1 | - | 10 - 11 | 1384 | 61 | |
More Information
Set Membership: |
|

PPP1R16B
Genomic Map