Assay Details
| Gene Symbol: NDUFA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175718.2 | NP_783649.1 | 2 - 3 | 244 | 61 | |
| GenBank mRNA | AJ510148.1 | - | 2 - 3 | 244 | 61 | |
| BC102976.1 | - | 2 - 3 | 265 | 61 | ||
More Information
Set Membership: |
|

NDUFA11
Genomic Map