Assay Details
| Gene Symbol: COX7A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175807.1 | NP_787001.1 | 2 - 3 | 181 | 81 | |
| GenBank mRNA | BC102664.1 | - | 2 - 3 | 202 | 81 | |
| MK423994.1 | - | 2 - 3 | 113 | 81 | ||
| X15235.1 | - | 142 | 81 | |||
More Information
Set Membership: |
|

COX7A2
Genomic Map