Assay Details
| Gene Symbol: RTN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113260.1 | NP_001106731.1 | 8 - 9 | 1284 | 66 | |
| NM_182656.1 | NP_872597.1 | 5 - 6 | 549 | 66 | ||
| GenBank mRNA | BC120363.1 | - | 8 - 9 | 1284 | 66 | |
More Information
Set Membership: |
|

RTN2
Genomic Map