Assay Details
| Gene Symbol: AIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183082.2 | NP_898905.2 | 3 - 4 | 512 | 64 | |
| GenBank mRNA | AY339893.1 | - | 3 - 4 | 498 | 64 | |
| BC103018.1 | - | 3 - 4 | 499 | 64 | ||
More Information
Set Membership: |
|

AIP
Genomic Map