Assay Details
| Gene Symbol: RPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033613.2 | NP_001028785.1 | 4 - 5 | 400 | 86 | |
| GenBank mRNA | AB098951.1 | - | 3 - 4 | 309 | 86 | |
| AF013215.1 | - | 3 - 4 | 356 | 86 | ||
| BC102227.1 | - | 4 - 5 | 402 | 86 | ||
More Information
Set Membership: |
|

RPS2
Genomic Map