Assay Details
| Gene Symbol: RPS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033624.2 | NP_001028796.1 | 2 - 3 | 208 | 51 | |
| GenBank mRNA | BC102215.1 | - | 2 - 3 | 209 | 51 | |
More Information
Set Membership: |
|

RPS16
Genomic Map