Assay Details
| Gene Symbol: GDI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033762.2 | NP_001028934.1 | 9 - 10 | 1372 | 60 | |
| GenBank mRNA | BC105197.1 | - | 9 - 10 | 1373 | 60 | |
| BT030504.1 | - | 9 - 10 | 1357 | 60 | ||
More Information
Set Membership: |
|

GDI2
Genomic Map