Assay Details
| Gene Symbol: CFH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033936.1 | NP_001029108.1 | 2 - 3 | 291 | 89 | |
| GenBank mRNA | BC105258.1 | - | 2 - 3 | 291 | 89 | |
More Information
Set Membership: |
|

CFH
Genomic Map