Assay Details
| Gene Symbol: COA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034256.2 | NP_001029428.1 | 3 - 4 | 208 | 114 | |
| GenBank mRNA | BC102530.1 | - | 3 - 4 | 209 | 114 | |
| BT026526.1 | - | 1 - 2 | 380 | 114 | ||
More Information
Set Membership: |
|

COA1
Genomic Map