Assay Details
| Gene Symbol: MPST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034291.2 | NP_001029463.2 | 2 - 3 | 722 | 118 | |
| NM_001244324.1 | NP_001231253.1 | 2 - 3 | 703 | 118 | ||
| GenBank mRNA | BC105248.1 | - | 2 - 3 | 689 | 118 | |
| EV680002.1 | - | 2 - 3 | 707 | 118 | ||
More Information
Set Membership: |
|

MPST
Genomic Map