Assay Details
| Gene Symbol: NOL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034384.2 | NP_001029556.1 | 7 - 8 | 729 | 84 | |
| GenBank mRNA | BC103151.1 | - | 7 - 8 | 730 | 84 | |
More Information
Set Membership: |
|

NOL7
Genomic Map