Assay Details
| Gene Symbol: PEBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034440.1 | NP_001029612.1 | 1 - 2 | 234 | 152 | |
| GenBank mRNA | BC102615.1 | - | 1 - 2 | 234 | 152 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102615.1, NM_001034440.1 transcripts.
Set Membership: |
|

PEBP4
Genomic Map