Assay Details
| Gene Symbol: AMZ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034481.1 | NP_001029653.1 | 6 - 7 | 954 | 60 | |
| GenBank mRNA | BC102726.1 | - | 6 - 7 | 954 | 60 | |
More Information
Set Membership: |
|

AMZ2
Genomic Map