Assay Details
| Gene Symbol: PUS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034512.2 | NP_001029684.1 | 2 - 3 | 987 | 141 | |
| GenBank mRNA | BC104569.1 | - | 2 - 3 | 992 | 141 | |
More Information
Set Membership: |
|

PUS3
Genomic Map