Assay Details
| Gene Symbol: SNAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034516.1 | NP_001029688.1 | 3 - 4 | 424 | 83 | |
| GenBank mRNA | BC102380.1 | - | 3 - 4 | 424 | 83 | |
More Information
Set Membership: |
|

SNAPC2
Genomic Map