Assay Details
| Gene Symbol: CYB5R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034518.1 | NP_001029690.1 | 3 - 4 | 278 | 58 | |
| GenBank mRNA | BC104584.1 | - | 3 - 4 | 278 | 58 | |
More Information
Set Membership: |
|

CYB5R1
Genomic Map