Assay Details
| Gene Symbol: C1QBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034527.2 | NP_001029699.1 | 2 - 3 | 493 | 93 | |
| GenBank mRNA | BC102464.1 | - | 2 - 3 | 494 | 93 | |
More Information
Set Membership: |
|

C1QBP
Genomic Map