Assay Details
| Gene Symbol: SFT2D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034551.2 | NP_001029723.1 | 7 - 8 | 511 | 72 | |
| GenBank mRNA | BC103331.1 | - | 7 - 8 | 513 | 72 | |
More Information
Set Membership: |
|

SFT2D1
Genomic Map