Assay Details
| Gene Symbol: S100PBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034687.2 | NP_001029859.1 | 5 - 6 | 1137 | 68 | |
| GenBank mRNA | BC105238.1 | - | 5 - 6 | 1138 | 68 | |
More Information
Set Membership: |
|

S100PBP
Genomic Map