Assay Details
| Gene Symbol: U2AF1L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034778.2 | NP_001029950.1 | 7 - 8 | 708 | 64 | |
| GenBank mRNA | BC102151.1 | - | 7 - 8 | 709 | 64 | |
More Information
Set Membership: |
|

U2AF1L4
Genomic Map