Assay Details
| Gene Symbol: PRR15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034794.2 | NP_001029966.1 | 1 - 2 | 228 | 104 | |
| GenBank mRNA | BC102073.1 | - | 1 - 2 | 230 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102073.1, NM_001034794.2 transcripts.
Set Membership: |
|

PRR15L
Genomic Map