Assay Details
| Gene Symbol: CEBPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034801.1 | NP_001029973.1 | 1 - 2 | 100 | 84 | |
| GenBank mRNA | BC102461.1 | - | 1 - 2 | 100 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102461.1, NM_001034801.1 transcripts.
Set Membership: |
|

CEBPG
Genomic Map