Assay Details
| Gene Symbol: MYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035025.2 | NP_001030197.1 | 2 - 3 | 133 | 67 | |
| GenBank mRNA | BC102914.1 | - | 2 - 3 | 134 | 67 | |
More Information
Set Membership: |
|

MYL2
Genomic Map