Assay Details
| Gene Symbol: FSCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035045.1 | NP_001030217.1 | 4 - 5 | 1409 | 56 | |
| GenBank mRNA | BC105199.1 | - | 4 - 5 | 1409 | 56 | |
More Information
Set Membership: |
|

FSCN1
Genomic Map