Assay Details
| Gene Symbol: RAB7A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035081.1 | NP_001030253.1 | 4 - 5 | 480 | 59 | |
| GenBank mRNA | BC102415.1 | - | 4 - 5 | 480 | 59 | |
More Information
Set Membership: |
|

RAB7A
Genomic Map