Assay Details
| Gene Symbol: ADPRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035340.2 | NP_001030417.1 | 5 - 6 | 823 | 76 | |
| GenBank mRNA | BC103360.1 | - | 5 - 6 | 824 | 76 | |
More Information
Set Membership: |
|

ADPRS
Genomic Map