Assay Details
| Gene Symbol: H1-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035372.1 | NP_001030449.1 | 4 - 5 | 797 | 100 | |
| GenBank mRNA | BC122822.1 | - | 3 - 4 | 491 | 100 | |
| DQ206443.1 | - | 4 - 5 | 797 | 100 | ||
More Information
Set Membership: |
|

H1-8
Genomic Map