Assay Details
| Gene Symbol: RAB11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035391.2 | NP_001030468.1 | 3 - 4 | 486 | 67 | |
| GenBank mRNA | BC105162.1 | - | 3 - 4 | 488 | 67 | |
More Information
Set Membership: |
|

RAB11B
Genomic Map