Assay Details
| Gene Symbol: HSD17B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035395.2 | NP_001030472.1 | 1 - 2 | 42 | 63 | |
| GenBank mRNA | BC102626.1 | - | 1 - 2 | 43 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102626.1, NM_001035395.2 transcripts.
Set Membership: |
|

HSD17B6
Genomic Map