Assay Details
| Gene Symbol: HSD17B6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035395.2 | NP_001030472.1 | 3 - 4 | 630 | 70 | |
| GenBank mRNA | BC102626.1 | - | 3 - 4 | 631 | 70 | |
More Information
Set Membership: |
|

HSD17B6
Genomic Map