Assay Details
| Gene Symbol: NSMCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035406.1 | NP_001030483.1 | 5 - 6 | 528 | 62 | |
| GenBank mRNA | BC102214.1 | - | 5 - 6 | 528 | 62 | |
More Information
Set Membership: |
|

NSMCE1
Genomic Map