Assay Details
| Gene Symbol: SMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035471.1 | NP_001030548.1 | 2 - 3 | 283 | 79 | |
| GenBank mRNA | BC103012.1 | - | 2 - 3 | 283 | 79 | |
| BC151416.1 | - | 2 - 3 | 209 | 79 | ||
| BT029883.1 | - | 2 - 3 | 236 | 79 | ||
More Information
Set Membership: |
|

SMS
Genomic Map