Assay Details
| Gene Symbol: TRMT10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035477.2 | NP_001030554.1 | 3 - 4 | 512 | 72 | |
| GenBank mRNA | BC105223.1 | - | 3 - 4 | 513 | 72 | |
More Information
Set Membership: |
|

TRMT10A
Genomic Map