Assay Details
| Gene Symbol: RANGRF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037460.2 | NP_001032537.1 | 1 - 2 | 200 | 90 | |
| GenBank mRNA | BC108151.1 | - | 1 - 2 | 201 | 90 | |
More Information
Set Membership: |
|

RANGRF
Genomic Map