Assay Details
| Gene Symbol: OTOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037484.2 | NP_001032561.1 | 3 - 4 | 407 | 89 | |
| GenBank mRNA | BC108168.1 | - | 3 - 4 | 408 | 89 | |
More Information
Set Membership: |
|

OTOR
Genomic Map