Assay Details
| Gene Symbol: LSM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037487.2 | NP_001032564.1 | 2 - 3 | 142 | 93 | |
| GenBank mRNA | BC108142.1 | - | 2 - 3 | 143 | 93 | |
More Information
Set Membership: |
|

LSM3
Genomic Map