Assay Details
| Gene Symbol: MRPS33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037605.2 | NP_001032694.1 | 3 - 4 | 333 | 104 | |
| GenBank mRNA | BC109522.1 | - | 3 - 4 | 334 | 104 | |
More Information
Set Membership: |
|

MRPS33
Genomic Map