Assay Details
| Gene Symbol: UBL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037710.2 | NP_001032799.1 | 2 - 3 | 245 | 62 | |
| GenBank mRNA | BC102198.1 | - | 2 - 3 | 246 | 62 | |
More Information
Set Membership: |
|

UBL5
Genomic Map