Assay Details
| Gene Symbol: IPO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001372122.1 | NP_001359051.1 | 12 - 13 | 1253 | 62 | |
| GenBank mRNA | BC107531.1 | - | 9 - 10 | 1030 | 62 | |
More Information
Set Membership: |
|

IPO5
Genomic Map