Assay Details
| Gene Symbol: PCNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038205.1 | NP_001033294.1 | 3 - 4 | 363 | 74 | |
| GenBank mRNA | BC108137.1 | - | 3 - 4 | 363 | 74 | |
More Information
Set Membership: |
|

PCNP
Genomic Map