Assay Details
| Gene Symbol: TRAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040476.3 | NP_001035566.2 | 10 - 11 | 1255 | 79 | |
| GenBank mRNA | BC104557.1 | - | 10 - 11 | 1145 | 79 | |
| EH201032.1 | - | 1 - 2 | 49 | 79 | ||
| GH306836.1 | - | 5 - 6 | 508 | 79 | ||
More Information
Set Membership: |
|

TRAM1
Genomic Map