Assay Details
| Gene Symbol: RPL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040516.2 | NP_001035606.1 | 4 - 5 | 390 | 82 | |
| GenBank mRNA | BC102223.1 | - | 4 - 5 | 391 | 82 | |
| BC142030.1 | - | 4 - 5 | 388 | 82 | ||
More Information
Set Membership: |
|

RPL19
Genomic Map